Acetimidohydrazide hydrochloride 95

Supplier: ALADDIN SCIENTIFIC
Warning

A193172-100MG A193172-1G A193172-250MG A193172-25MG A193172-5G
ALA193172100MGEA 104.41 USD
ALA193172100MG ALA193172-1G ALA193172250MG ALA193172-25MG ALA193172-5G
Acetimidohydrazide hydrochloride 95
Acetimidohydrazide hydrochloride
Formula: C₂H₈ClN₃
MW: 109.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01746339
CAS Number: 39254-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725fda7e8b953dc26713d19fec0a0557eee10031&vid=725fda7e8b953dc26713d19fec0a0557eee10031&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR