2-Amino-6-chlorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A193106-250MG A193106-5G A193106-1G
ALA193106250MGEA 10.36 USD
ALA193106250MG ALA193106-5G ALA193106-1G
2-Amino-6-chlorophenol ≥98%
2-Amino-6-chlorophenol

Application:2-Amino-6-Chlorophenol is used to synthesize HT2C receptor agonists with high selectivity in functional and binding assays.

Formula: C₆H₆ClNO
Boiling Pt: 247℃
Melting Pt: 80-81 ºC
Density: 1.406 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD07801210
CAS Number: 38191-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11c4749e1b231dcbc6b651f41e971de9fa733f9a&vid=11c4749e1b231dcbc6b651f41e971de9fa733f9a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR