(Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(3-(3-(tert-butyl)-4-hydroxy-5-methylphenyl)propanoate) ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Antioxidant 245, [Ethane-1,2-diylbis(oxy)]bis(ethane-2,1-diyl) Bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate], Triethylene glycol bis(3-tert-butyl-4-hydroxy-5-methylphenyl)propionate, Irganox 245

A193004-100G A193004-1G A193004-25G A193004-5G
ALA193004-100GEA 23.66 USD
ALA193004-100G ALA193004-1G ALA193004-25G ALA193004-5G
(Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(3-(3-(tert-butyl)-4-hydroxy-5-methylphenyl)propanoate) ≥98%
(Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(3-(3-(tert-butyl)-4-hydroxy-5-methylphenyl)propanoate)
Formula: C₃₄H₅₀O₈
MW: 586.75 g/mol
Melting Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134694
CAS Number: 36443-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad707a31a14d1aaf70e3b140ddfa2d57ca4d9223&vid=ad707a31a14d1aaf70e3b140ddfa2d57ca4d9223&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR