2-Amino-6-chlorobenzaldehyde ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A192956-100MG A192956-10G A192956-1G A192956-250MG A192956-5G
ALA192956100MGEA 27.46 USD
ALA192956100MG ALA192956-10G ALA192956-1G ALA192956250MG ALA192956-5G
2-Amino-6-chlorobenzaldehyde ≥95%
2-Amino-6-chlorobenzaldehyde
Formula: C₇H₆ClNO
MW: 155.582 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696880
CAS Number: 35490-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725bb965281ed42aceb5d3113f8e57fcb984cf79&vid=725bb965281ed42aceb5d3113f8e57fcb984cf79&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR