2-Aminododecanoic acid 97

Supplier: ALADDIN SCIENTIFIC
Warning

A192935-1G A192935-250MG A192935-5G
ALA192935-1GEA 130.06 USD
ALA192935-1G ALA192935250MG ALA192935-5G
2-Aminododecanoic acid 97
2-Aminododecanoic acid
Formula: C₁₂H₂₅NO₂
MW: 215.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020390
CAS Number: 35237-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cbbbc2822a4e416a3039434a376825294f25988&vid=1cbbbc2822a4e416a3039434a376825294f25988&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR