(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-2-Fluoro-5-Nitrobenzophenone

A192869-1G A192869-25G A192869-5G A192869-100G
ALA192869-1GEA 9.41 USD
ALA192869-1G ALA192869-25G ALA192869-5G ALA192869-100G
(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone ≥98%
(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone

Application:Intermediate in the production of Flunitrazepam metabolites.

Formula: C₁₃H₉FN₂O₃
MW: 260.22 g/mol
Melting Pt: 158-160°C
MDL Number: MFCD00970341
CAS Number: 344-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=178af569e3c34763ffd74d22a8761065ef43387d&vid=178af569e3c34763ffd74d22a8761065ef43387d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR