5-Amino-1H-pyrazol-3(2H)-one ≥98%

Supplier: Aladdin Scientific
Warning

A192525-100G A192525-1G A192525-25G A192525-5G
ALA192525-100GEA 212.71 USD
ALA192525-100G ALA192525-1G ALA192525-25G ALA192525-5G
5-Amino-1H-pyrazol-3(2H)-one ≥98%
5-Amino-1H-pyrazol-3(2H)-one
Formula: C₃H₅N₃O
MDL Number: MFCD00204482
CAS Number: 28491-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc9a80088d1c7a1e16774232ac46d68014a8045&vid=5cc9a80088d1c7a1e16774232ac46d68014a8045&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR