Ammonium dimolybdate Mo 56.5%

Supplier: ALADDIN SCIENTIFIC

A192483-100G A192483-25G
ALA192483-100GEA 85.41 USD
ALA192483-100G ALA192483-25G
Ammonium dimolybdate Mo 56.5%
Ammonium dimolybdate
Formula: H₈Mo₂N₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00010882
CAS Number: 27546-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0434474ee41e734dbefc7be4dfb43905fdc22e1&vid=c0434474ee41e734dbefc7be4dfb43905fdc22e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR