3-Aminopyrazine-2-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A192366-1G A192366-250MG A192366-25G A192366-5G
ALA192366-1GEA 50.26 USD
ALA192366-1G ALA192366250MG ALA192366-25G ALA192366-5G
3-Aminopyrazine-2-carbonitrile ≥98%
3-Aminopyrazine-2-carbonitrile
Formula: C5H4N4
MW: 120.1121 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02091510
CAS Number: 25911-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ca2f77dc523678e7e290f9d551813d9ce95ea7&vid=f4ca2f77dc523678e7e290f9d551813d9ce95ea7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR