1-(5-Aminopentyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A192133-100MG A192133-1G A192133-25MG
ALA192133100MGEA 96.81 USD
ALA192133100MG ALA192133-1G ALA192133-25MG
1-(5-Aminopentyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate ≥95%
1-(5-Aminopentyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate
Formula: C₁₁H₁₅F₃N₂O₄
MW: 296.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01073616
CAS Number: 222159-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf2d5c06c173489b91154cedc99634dcfe93a086&vid=cf2d5c06c173489b91154cedc99634dcfe93a086&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR