(2-Amino-3-methoxyphenyl)methanol 95
Supplier: ALADDIN SCIENTIFIC
Warning
|
A191970-100MG
A191970-1G
A191970-250MG
A191970-25MG
A191970-5G
ALA191970100MGEA
47.41
USD
ALA191970100MG
ALA191970-1G
ALA191970250MG
ALA191970-25MG
ALA191970-5G
(2-Amino-3-methoxyphenyl)methanol 95
(2-Amino-3-methoxyphenyl)methanol
Formula:
C₈H₁₁NO₂ MW: 153.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09029933 CAS Number: 205877-13-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...