4-Fluorobenzo[d]thiazol-2-amine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4-fluorobenzothiazole

A191942-1G A191942-25G
ALA191942-1GEA 13.21 USD
ALA191942-1G ALA191942-25G
4-Fluorobenzo[d]thiazol-2-amine ≥97%
4-Fluorobenzo[d]thiazol-2-amine
Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04448824
CAS Number: 20358-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1d57e17c4aa9038ed35091b1d107f6d400b08c2&vid=d1d57e17c4aa9038ed35091b1d107f6d400b08c2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR