2-Amino-5-chlorobenzothiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A191940-10G A191940-1G A191940-25G A191940-5G
ALA191940-10GEA 310.56 USD
ALA191940-10G ALA191940-1G ALA191940-25G ALA191940-5G
2-Amino-5-chlorobenzothiazole ≥97%
2-Amino-5-chlorobenzothiazole
Formula: C₇H₅ClN₂S
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00205355
CAS Number: 20358-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21df65ae390ce74f29d1b681669f0f20ae55f9f&vid=b21df65ae390ce74f29d1b681669f0f20ae55f9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR