[2-(4-Aminomethylphenoxy)ethyl]dimethylamine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
2-(4-(Aminomethyl)phenoxy)-N,N-dimethylethanamine, 2-[4-(Amino methyl) phenoxy]-n, n-dimethyl ethanamine, Trimethobenzamide Impurity 1
A191907-100G
A191907-1G
A191907-250MG
A191907-25G
A191907-5G
ALA191907-100GEA
1094.31
USD
ALA191907-100G
ALA191907-1G
ALA191907250MG
ALA191907-25G
ALA191907-5G
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine ≥97%
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine
4-[2-(Dimethylamino)ethoxy]benzylamine is an impurity of Itopride (HCl: I931500), a gastroprokinetic benzamide derivative that is used to treat patients with dyspepsia.
Formula:
C₁₁H₁₈N₂O MW: 194.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01075231 CAS Number: 20059-73-8 EINECS: 243-491-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...