4-Amino-3-chlorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A191581-100G A191581-1G A191581-25G A191581-5G
ALA191581-100GEA 15809.81 USD
ALA191581-100G ALA191581-1G ALA191581-25G ALA191581-5G
4-Amino-3-chlorophenol ≥98%
4-Amino-3-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00801116
CAS Number: 17609-80-2
EINECS: 241-583-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e684962cf8b17b7b771e454fe77d6b219a11f9c&vid=7e684962cf8b17b7b771e454fe77d6b219a11f9c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR