5-Amino-2-chloro-4-nitrobenzotrifluoride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Nitro-4-chloro-5-(trifluoromethyl)aniline, 5-Amino-2-chloro-4-nitrobenzotrifluoride

A191423-100G A191423-1G A191423-250MG A191423-25G A191423-5G
ALA191423-100GEA 1081.01 USD
ALA191423-100G ALA191423-1G ALA191423250MG ALA191423-25G ALA191423-5G
5-Amino-2-chloro-4-nitrobenzotrifluoride ≥95%
5-Amino-2-chloro-4-nitrobenzotrifluoride
Formula: C₇H₄ClF₃N₂O₂
MDL Number: MFCD03094264
CAS Number: 167415-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d197ca5dc75678cc000f41107c4d7e9cb2abff&vid=90d197ca5dc75678cc000f41107c4d7e9cb2abff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR