Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Allyl 3,5-diamino-1H-pyrazole-4-carboxylate, 3,5-diamino-1H-pyrazole-4-carboxylate

A191264-1G A191264-250MG A191264-25G A191264-5G
ALA191264-1GEA 164.26 USD
ALA191264-1G ALA191264250MG ALA191264-25G ALA191264-5G
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate ≥98%
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Formula: C₇H₁₀N₄O₂
MW: 182.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27996731
CAS Number: 1613191-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e0d5b05da75509b946c8bdc4926a87060c42369&vid=5e0d5b05da75509b946c8bdc4926a87060c42369&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR