2-Amino-5-bromobenzoylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (2-Amino-5-bromophenyl)(pyridin-2-yl)methanone, 2-(2-Amino-5-bromobenzoyl)pyridine

A191151-25G A191151-5G A191151-100G A191151-1G
ALA191151-25GEA 138.61 USD
ALA191151-25G ALA191151-5G ALA191151-100G ALA191151-1G
2-Amino-5-bromobenzoylpyridine ≥98%
2-Amino-5-bromobenzoylpyridine

Product Application:Intermediate in the preparation of Bromazepam.

Formula: C₁₂H₉BrN₂O
MW: 277.1167 g/mol
Melting Pt: 98 - 100°C
Storage Temperature: Refrigerator
MDL Number: MFCD07780283
CAS Number: 1563-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2c79babad25b9328de5816cc0e3c37afbb641f6&vid=a2c79babad25b9328de5816cc0e3c37afbb641f6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR