4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one ≥97%

Supplier: Aladdin Scientific
Danger

A191108-1G A191108-250MG A191108-5G
ALA191108-1GEA 10.36 USD
ALA191108-1G ALA191108250MG ALA191108-5G
4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one ≥97%
4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one
Formula: C₁₁H₁₆O₂
MDL Number: MFCD06409997
CAS Number: 15356-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df5866571127a1e1c591982be9d4bfcb04b359a7&vid=df5866571127a1e1c591982be9d4bfcb04b359a7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR