2-Amino-3-bromobenzaldehyde ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A190962-1G A190962-50MG A190962-5G
ALA190962-1GEA 52.16 USD
ALA190962-1G ALA190962-50MG ALA190962-5G
2-Amino-3-bromobenzaldehyde ≥98%
2-Amino-3-bromobenzaldehyde
Formula: C₇H₆BrNO
MDL Number: MFCD18389339
CAS Number: 145123-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bea7d9f1b9be64f03d711f110330f1db504ae32&vid=7bea7d9f1b9be64f03d711f110330f1db504ae32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR