2-Chloro-DL-phenylalanine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Chloro-DL-phenylalanine, 2-Amino-3-(2-chlorophenyl)propanoic acid, 2-Chlorophenylalanine

A190846-1G A190846-250MG A190846-25G A190846-100G
ALA190846-1GEA 15.11 USD
ALA190846-1G ALA190846250MG ALA190846-25G ALA190846-100G
2-Chloro-DL-phenylalanine ≥97%
2-Chloro-DL-phenylalanine
Formula: C₉H₁₀ClNO₂
MW: 199.6300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037774
CAS Number: 14091-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46732ed8ec513eb2824847f629148f6726234f2f&vid=46732ed8ec513eb2824847f629148f6726234f2f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR