1-(2-Amino-4-bromophenyl)ethanone ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A190249-10G
A190249-1G
A190249-250MG
A190249-50MG
A190249-5G
ALA190249-10GEA
313.41
USD
ALA190249-10G
ALA190249-1G
ALA190249250MG
ALA190249-50MG
ALA190249-5G
1-(2-Amino-4-bromophenyl)ethanone ≥98%
1-(2-Amino-4-bromophenyl)ethanone
Formula:
C₈H₈BrNO MW: 214.05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16659002 CAS Number: 123858-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...