2-(4-Aminobenzyl)aniline ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-[(4-aminophenyl)methyl]aniline

A190082-100MG A190082-1G A190082-250MG A190082-25MG A190082-5G
ALA190082100MGEA 245.96 USD
ALA190082100MG ALA190082-1G ALA190082250MG ALA190082-25MG ALA190082-5G
2-(4-Aminobenzyl)aniline ≥95%
2-(4-Aminobenzyl)aniline
Formula: C₁₃H₁₄N₂
MW: 198.264 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01670227
CAS Number: 1208-52-2
ADR: 6.1,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5994979d92fdd929f39ed88716102f9d46d3aa8c&vid=5994979d92fdd929f39ed88716102f9d46d3aa8c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR