3-Amino-3-(4-isopropylphenyl)propionic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A189895-1G A189895-250MG A189895-25G A189895-5G
ALA189895-1GEA 32.21 USD
ALA189895-1G ALA189895250MG ALA189895-25G ALA189895-5G
3-Amino-3-(4-isopropylphenyl)propionic acid ≥97%
3-Amino-3-(4-isopropylphenyl)propionic acid
Formula: C₁₂H₁₇NO₂
MW: 207.2700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663184
CAS Number: 117391-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d884e8c0504b52839fa4e443696b3dad0ea8024e&vid=d884e8c0504b52839fa4e443696b3dad0ea8024e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR