Diammonium oxalate ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A189700-100G
A189700-10G
A189700-25G
A189700-500G
A189700-50G
ALA189700-100GEA
19.86
USD
ALA189700-100G
ALA189700-10G
ALA189700-25G
ALA189700-500G
ALA189700-50G
Diammonium oxalate ≥98%
Diammonium oxalate
Formula:
C₂H₈N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00013308 CAS Number: 1113-38-8 UN: 1759 ADR: 8,I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...