α-Bromostyrene ≥95% stabilized

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: alfa-Bromostyrene, α-Bromostyrene, alpha-Bromostyrene

A189239-100G A189239-1G A189239-25G A189239-5G
ALA189239-100GEA 747.56 USD
ALA189239-100G ALA189239-1G ALA189239-25G ALA189239-5G
α-Bromostyrene ≥95% stabilized
α-Bromostyrene
Formula: C₈H₇Br
MW: 183.0452 g/mol
Density: 1.41 g/cm³
Flash Pt: 86°C
Storage Temperature: Freezer
MDL Number: MFCD00012229
CAS Number: 98-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1722b07c062c6dc9d50f7df93dd7ab3a52fb9869&vid=1722b07c062c6dc9d50f7df93dd7ab3a52fb9869&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR