3-Amino-2-fluorobenzoic acid ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A189229-1G
A189229-25G
A189229-5G
ALA189229-1GEA
21.76
USD
ALA189229-1G
ALA189229-25G
ALA189229-5G
3-Amino-2-fluorobenzoic acid ≥98%
3-Amino-2-fluorobenzoic acid
Formula:
C7H6FNO2 MW: 155.1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797827 CAS Number: 914223-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...