5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A189203-25G A189203-5G
ALA189203-25GEA 34.11 USD
ALA189203-25G ALA189203-5G
5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97%
5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
Formula: C₁₄H₁₈I₃N₃O₆
Melting Pt: 195°C
Storage Temperature: Ambient
MDL Number: MFCD08063355
CAS Number: 76801-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ee04ddb63f759d4d67c2b50d8b7b99599a05a99&vid=9ee04ddb63f759d4d67c2b50d8b7b99599a05a99&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR