Ammonium polyphosphate n≥1000
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Polyphosphoric acids ammonium salts
A189185-100G
A189185-25G
ALA189185-100GEA
24.61
USD
ALA189185-100G
ALA189185-25G
Ammonium polyphosphate n≥1000
Ammonium polyphosphate
Formula:
(NH₄PO₃)n Storage Temperature: Ambient |
CAS Number:
68333-79-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...