Sodium 4-(3-methyl-5-oxo-4-(phenyldiazenyl)-4,5-dihydro-1H-pyrazol-1-yl)benzenesulfonate biological stain

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: acid yellow 11

A189176-100G A189176-25G
ALA189176-100GEA 159.51 USD
ALA189176-100G ALA189176-25G
Sodium 4-(3-methyl-5-oxo-4-(phenyldiazenyl)-4,5-dihydro-1H-pyrazol-1-yl)benzenesulfonate biological stain
Sodium 4-(3-methyl-5-oxo-4-(phenyldiazenyl)-4,5-dihydro-1H-pyrazol-1-yl)benzenesulphonate
Formula: C₁₆H₁₃N₄NaO₄S
MW: 380.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003122
CAS Number: 6359-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c546eae1294ce3e82fa1b4ac620e62c68dbefac&vid=7c546eae1294ce3e82fa1b4ac620e62c68dbefac&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR