Allyl Maleate ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A189086-1G A189086-25G A189086-5G
ALA189086-1GEA 23.66 USD
ALA189086-1G ALA189086-25G ALA189086-5G
Allyl Maleate ≥95%
Allyl Maleate
Formula: C₇H₈O₄
Storage Temperature: Refrigerator
CAS Number: 2424-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=564f4ec2b3a96fb71962ac4fe4d8bf6555fd657d&vid=564f4ec2b3a96fb71962ac4fe4d8bf6555fd657d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR