Ammonium magnesium phosphate hexahydrate ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A189001-100G
A189001-25G
A189001-5G
ALA189001-100GEA
67.36
USD
ALA189001-100G
ALA189001-25G
ALA189001-5G
Ammonium magnesium phosphate hexahydrate ≥98%
Ammonium magnesium phosphate hexahydrate
Formula:
NH₄MgPO₄·6H₂O MW: 245.41 g/mol (137.32 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150461 CAS Number: 13478-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...