2-Amino-5-cyanobenzoic acid ≥96%

Supplier: ALADDIN SCIENTIFIC

A188963-1G A188963-250MG A188963-5G
ALA188963-1GEA 109.16 USD
ALA188963-1G ALA188963250MG ALA188963-5G
2-Amino-5-cyanobenzoic acid ≥96%
2-Amino-5-cyanobenzoic acid
Formula: C₈H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20485816
CAS Number: 99767-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae120cee5e6b8e9fa25c0f48d9015c7a62ce0d1c&vid=ae120cee5e6b8e9fa25c0f48d9015c7a62ce0d1c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR