6-Morpholinopyrimidin-4-amine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Morpholinopyrimidin-4-amine, 4-Amino-6-morpholinopyrimidine

A188889-1G A188889-250MG A188889-5G
ALA188889-1GEA 59.76 USD
ALA188889-1G ALA188889250MG ALA188889-5G
6-Morpholinopyrimidin-4-amine ≥98%
6-Morpholinopyrimidin-4-amine
Formula: C₈H₁₂N₄O
MW: 180.2071 g/mol
MDL Number: MFCD09999228
CAS Number: 96225-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef4bfc123f23195f45b61e643a92a532ea10d900&vid=ef4bfc123f23195f45b61e643a92a532ea10d900&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR