2-(4-Amino-1H-pyrazol-1-yl)ethanol ≥95%

Supplier: ALADDIN SCIENTIFIC

A188563-1G A188563-250MG A188563-50MG A188563-5G
ALA188563-1GEA 127.21 USD
ALA188563-1G ALA188563250MG ALA188563-50MG ALA188563-5G
2-(4-Amino-1H-pyrazol-1-yl)ethanol ≥95%
2-(4-Amino-1H-pyrazol-1-yl)ethanol
Formula: C₅H₉N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD11147853
CAS Number: 948571-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73ccec3df2f5718f2a43e5413fbca57c3ab16fe8&vid=73ccec3df2f5718f2a43e5413fbca57c3ab16fe8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR