2-Aminobenzo[d]thiazole-6-carboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Aminobenzothiazole-6-carboxylic acid

A188429-25G A188429-5G
ALA188429-25GEA 820.71 USD
ALA188429-25G ALA188429-5G
2-Aminobenzo[d]thiazole-6-carboxylic acid ≥98%
2-Aminobenzo[d]thiazole-6-carboxylic acid
Formula: C₈H₆N₂O₂S
MDL Number: MFCD00054180
CAS Number: 93-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a019c73ed659b6aef0d260686bf5f235594709c&vid=3a019c73ed659b6aef0d260686bf5f235594709c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR