5-Aminoisoquinolin-1(2H)-one ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A188364-100MG A188364-1G A188364-250MG A188364-5G
ALA188364100MGEA 84.46 USD
ALA188364100MG ALA188364-1G ALA188364250MG ALA188364-5G
5-Aminoisoquinolin-1(2H)-one ≥98%
5-Aminoisoquinolin-1(2H)-one
Formula: C₉H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06198896
CAS Number: 93117-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb2416bf16eec08177325b02d3acc9f37f3d40a5&vid=cb2416bf16eec08177325b02d3acc9f37f3d40a5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR