3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-(2-Amino-5-bromo-pyridin-3-yl)-acrylic acid methyl ester

A188061-1G A188061-250MG A188061-50MG
ALA188061-1GEA 1781.16 USD
ALA188061-1G ALA188061250MG ALA188061-50MG
3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester ≥95%
3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester
Formula: C₉H₉BrN₂O₂
MW: 257.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781129
CAS Number: 912760-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbf737b8640cfd15d137fe7187061d4d115f6630&vid=cbf737b8640cfd15d137fe7187061d4d115f6630&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR