6-Methoxyquinolin-8-amine ≥96%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A188001-100G
A188001-1G
A188001-25G
A188001-5G
ALA188001-100GEA
1001.21
USD
ALA188001-100G
ALA188001-1G
ALA188001-25G
ALA188001-5G
6-Methoxyquinolin-8-amine ≥96%
6-Methoxyquinolin-8-amine
Formula:
C₁₀H₁₀N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00672902 CAS Number: 90-52-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...