4-Hydroxybenzylamine hydrobromide ≥97%

Supplier: ALADDIN SCIENTIFIC

A187992-25G A187992-5G
ALA187992-25GEA 201.31 USD
ALA187992-25G ALA187992-5G
4-Hydroxybenzylamine hydrobromide ≥97%
4-Hydroxybenzylamine hydrobromide
Formula: C₇H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD02258769
CAS Number: 90430-14-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e779fb56ea5f77669cad4c4d44a3a4e15df46f95&vid=e779fb56ea5f77669cad4c4d44a3a4e15df46f95&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR