2-Aminoethanesulfonamide hydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Aminoethanesulphonamide, HCl
A187897-1G
A187897-100G
ALA187897-1GEA
55.96
USD
ALA187897-1G
ALA187897-100G
2-Aminoethanesulfonamide hydrochloride ≥98%
2-Aminoethanesulfonamide hydrochloride
Formula:
C₂H₉ClN₂O₂S MW: 160.62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02258341 CAS Number: 89756-60-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...