Methyl-4-aminothiophene-2-carboxylate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-thiophene-2-carboxylic acid methyl ester, Methyl 4-aminothiophene-2-carboxylate

A187877-1G A187877-250MG A187877-25G A187877-5G
ALA187877-1GEA 84.46 USD
ALA187877-1G ALA187877250MG ALA187877-25G ALA187877-5G
Methyl-4-aminothiophene-2-carboxylate ≥98%
Methyl-4-aminothiophene-2-carboxylate
Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368326
CAS Number: 89499-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03d0b9bc76269bcd44d3c5593a1e1d4801d6af&vid=de03d0b9bc76269bcd44d3c5593a1e1d4801d6af&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR