2-Amino-5-iodo-4-methoxypyrimidine ≥95%
Supplier: Aladdin Scientific
Warning
|
A187850-1G
A187850-250MG
A187850-50MG
A187850-5G
ALA187850-1GEA
136.71
USD
ALA187850-1G
ALA187850250MG
ALA187850-50MG
ALA187850-5G
2-Amino-5-iodo-4-methoxypyrimidine ≥95%
2-Amino-5-iodo-4-methoxypyrimidine
Formula:
C₅H₆IN₃O Storage Temperature: Refrigerator |
MDL Number:
MFCD11520889 CAS Number: 89322-66-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...