3-(Aminomethyl)benzoic acid hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-(Aminomethyl)benzoic acid, HCl

A187526-1G A187526-250MG A187526-5G
ALA187526-1GEA 59.76 USD
ALA187526-1G ALA187526250MG ALA187526-5G
3-(Aminomethyl)benzoic acid hydrochloride ≥98%
3-(Aminomethyl)benzoic acid hydrochloride
Formula: C₈H₁₀ClNO₂
MW: 187.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03791117
CAS Number: 876-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca3179c50e9a0f34dc544cb923ac76641dc20543&vid=ca3179c50e9a0f34dc544cb923ac76641dc20543&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR