4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine hydrochloride, 4-Aminomethylphenylboronic acid pinacol ester HCl

A187049-1G A187049-250MG A187049-5G
ALA187049-1GEA 98.71 USD
ALA187049-1G ALA187049250MG ALA187049-5G
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride ≥95%
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Formula: C₁₃H₂₀BNO₂·HCl
Melting Pt: 228-238°C
Storage Temperature: Refrigerator
MDL Number: MFCD02179455
CAS Number: 850568-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=604a447f7a7ff9ec1cfe59b4e759840adf0863dd&vid=604a447f7a7ff9ec1cfe59b4e759840adf0863dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR