1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Acetyl-7(1H)-azaindole, 1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone
A186818-1G
A186818-25G
A186818-5G
ALA186818-1GEA
258.31
USD
ALA186818-1G
ALA186818-25G
ALA186818-5G
1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone ≥98%
1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone
Formula:
C₉H₈N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD09743457 CAS Number: 83393-46-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...