3-Amino-N-isopropylbenzamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A186750-100MG A186750-1G A186750-250MG A186750-5G
ALA186750100MGEA 13.21 USD
ALA186750100MG ALA186750-1G ALA186750250MG ALA186750-5G
3-Amino-N-isopropylbenzamide ≥97%
3-Amino-N-isopropylbenzamide
Formula: C₁₀H₁₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09045316
CAS Number: 81882-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed633fab088a75b62edafc532b7786a059e0307&vid=9ed633fab088a75b62edafc532b7786a059e0307&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR