(S)-3-Amino-4-oxo-4-(((R)-1-oxo-1-((2,2,4,4-tetramethylthietan-3-yl)amino)propan-2-yl)amino)butanoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC

A186704-1G A186704-250MG A186704-5G
ALA186704-1GEA 652.56 USD
ALA186704-1G ALA186704250MG ALA186704-5G
(S)-3-Amino-4-oxo-4-(((R)-1-oxo-1-((2,2,4,4-tetramethylthietan-3-yl)amino)propan-2-yl)amino)butanoic acid ≥95%
(S)-3-Amino-4-oxo-4-(((R)-1-oxo-1-((2,2,4,4-tetramethylthietan-3-yl)amino)propan-2-yl)amino)butanoic acid
Formula: C₁₄H₂₅N₃O₄S
Storage Temperature: Freezer
CAS Number: 80863-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cded231cd9d475d015b689d6b913511b177b0782&vid=cded231cd9d475d015b689d6b913511b177b0782&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR