2-Amino-3-bromo-5-(trifluoromethyl)pyridine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-3-bromo-5-(trifluoromethyl)pyridine, 3-Bromo-5-(trifluoromethyl)pyridin-2-amine, 3-Bromo-5-(trifluoromethyl)-2-pyridinylamine, 2-Amino-3-bromo-5-trifluoromethylpyridine
A186659-1G
A186659-250MG
A186659-25G
A186659-100G
ALA186659-1GEA
49.31
USD
ALA186659-1G
ALA186659250MG
ALA186659-25G
ALA186659-100G
2-Amino-3-bromo-5-(trifluoromethyl)pyridine ≥98%
2-Amino-3-bromo-5-(trifluoromethyl)pyridine
Formula:
C₆H₄BrF₃N₂ MW: 241.01 g/mol Melting Pt: 94-104°C Storage Temperature: Ambient |
MDL Number:
MFCD07375382 CAS Number: 79456-30-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...