2-Amino-4-(4-fluorophenyl)thiazole ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A186592-1G
A186592-250MG
A186592-25G
A186592-5G
ALA186592-1GEA
16.06
USD
ALA186592-1G
ALA186592250MG
ALA186592-25G
ALA186592-5G
2-Amino-4-(4-fluorophenyl)thiazole ≥97%
2-Amino-4-(4-fluorophenyl)thiazole
Formula:
C₉H₇FN₂S Melting Pt: 102-110°C Storage Temperature: Refrigerator |
MDL Number:
MFCD01086598 CAS Number: 77815-14-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...